Gene Transcripts

Gene Symbol
HSPA12B
Entrez Gene ID
116835
Gene Name
heat shock protein family A (Hsp70) member 12B
Gene Aliases
C20orf60
Chromosome Location
Chr. 20: 3732670 - 3753111 on Build GRCh38
UniGene ID
Hs.516854
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197327.1 NP_001184256.1 12 1486
NM_001318322.1 NP_001305251.1 11 1391
NM_052970.4 NP_443202.3 12 1489
XM_011529151.2 XP_011527453.1 9 1187
XM_017027632.1 XP_016883121.1 12 1404
XM_017027633.1 XP_016883122.1 12 1388
XM_017027634.1 XP_016883123.1 12 1388
XM_017027635.1 XP_016883124.1 12 1432
GenBank mRNA AK056712.1 - 12 1426
AK297120.1 - 10 1282
AK301294.1 - 11 1357
BC090857.1 - 11 1350
BC110881.1 - 12 1438 5725594
BC117284.1 - 12 1387 40125835
BC126263.1 - 12 1387 8991979
BC143932.1 - 12 1384 9052450

View Full Product Details