Gene Transcripts

Gene Symbol
OR4X2
Entrez Gene ID
119764
Gene Name
olfactory receptor family 4 subfamily X member 2 (gene/pseudogene)
Gene Aliases
OR11-105
Chromosome Location
Chr. 11: 48245104 - 48246015 on Build GRCh38
UniGene ID
Hs.553565
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004727.1 NP_001004727.1 1 835
GenBank mRNA BC136935.1 - 1 883
BC136936.1 - 1 883 9020928

View Full Product Details