Gene Transcripts

Gene Symbol
C16orf78
Entrez Gene ID
123970
Gene Name
chromosome 16 open reading frame 78
Gene Aliases
Chromosome Location
Chr. 16: 49373808 - 49399431 on Build GRCh38
UniGene ID
Hs.125875
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144602.3 NP_653203.1 2 301
GenBank mRNA BC021181.2 - 2 212
BC021718.2 - 2 212 5265370
HY001804.1 - 2 301

View Full Product Details