Gene Transcripts

Gene Symbol
TBC1D16
Entrez Gene ID
125058
Gene Name
TBC1 domain family member 16
Gene Aliases
Chromosome Location
Chr. 17: 79932343 - 80035875 on Build GRCh38
UniGene ID
Hs.369819
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271844.1 NP_001258773.1 5 630
NM_001271845.1 NP_001258774.1 5 669
NM_001271846.1 NP_001258775.1 5 669
NM_019020.3 NP_061893.2 9 1785
XM_005257049.2 XP_005257106.1 9 1796
XM_005257050.3 XP_005257107.1 9 1799
XM_005257052.3 XP_005257109.1 9 1802
XM_006721694.3 XP_006721757.1 9 1802
XM_006721695.3 XP_006721758.1 9 1802
XM_011524322.2 XP_011522624.1 9 1760
XM_011524323.2 XP_011522625.2 7 1710
XM_011524324.2 XP_011522626.1 5 1254
XM_017024185.1 XP_016879674.1 9 1799
XM_017024186.1 XP_016879675.1 7 1537
XM_017024187.1 XP_016879676.1 8 1777
XM_017024188.1 XP_016879677.1 7 917
GenBank mRNA AB449902.1 - 8 1659
BC001525.2 - 5 640
BC028290.1 - 9 1775 4480654
BC036947.1 - 5 674 5392579

View Full Product Details