Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153213.3 | NP_694945.2 | 2 | 391 | |
XM_011540706.2 | XP_011539008.1 | 3 | 555 | ||
GenBank mRNA | AK091281.1 | - | 2 | 427 | |
AK297707.1 | - | 2 | 453 | ||
BC040640.1 | - | 2 | 391 | 3847806 |