Gene Transcripts

Gene Symbol
RFTN2
Entrez Gene ID
130132
Gene Name
raftlin family member 2
Gene Aliases
C2orf11, Raftlin-2
Chromosome Location
Chr. 2: 197568224 - 197676050 on Build GRCh38
UniGene ID
Hs.591615
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144629.2 NP_653230.2 1 417
XM_011510595.2 XP_011508897.1 1 607
XM_011510596.2 XP_011508898.1 1 607
XM_011510597.2 XP_011508899.1 1 607
XM_011510598.2 XP_011508900.1 1 607
XM_017003332.1 XP_016858821.1 1 607
GenBank mRNA AK055136.1 - 1 417
BC093966.1 - 1 64 7939811
BC111069.1 - 1 549 5764096
BC113361.1 - 1 64 8322413

View Full Product Details