Gene Transcripts

Gene Symbol
SPATA4
Entrez Gene ID
132851
Gene Name
spermatogenesis associated 4
Gene Aliases
SPEF1B, TSARG2
Chromosome Location
Chr. 4: 176184574 - 176195671 on Build GRCh38
UniGene ID
Hs.481235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144644.3 NP_653245.2 5 825
GenBank mRNA AY040204.3 - 5 825
BC021731.2 - 5 726 5269042
BC039342.1 - 4 1793

View Full Product Details