Gene Transcripts

Gene Symbol
EVC2
Entrez Gene ID
132884
Gene Name
EvC ciliary complex subunit 2
Gene Aliases
LBN, WAD
Chromosome Location
Chr. 4: 5531921 - 5709548 on Build GRCh38
UniGene ID
Hs.87306
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166136.1 NP_001159608.1 3 896
NM_147127.4 NP_667338.3 3 458
XM_017007735.1 XP_016863224.1 3 469
XM_017007736.1 XP_016863225.1 4 447
XM_017007737.1 XP_016863226.1 4 334
XM_017007738.1 XP_016863227.1 3 469
XM_017007739.1 XP_016863228.1 4 500
GenBank mRNA AB083067.1 - 2 164
AY152402.1 - 3 893
AY152403.1 - 3 893
AY185210.1 - 3 458
BC143836.1 - 4 505

View Full Product Details