Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278305.1 | NP_001265234.1 | 10 | 668 | |
NM_001278306.1 | NP_001265235.1 | 9 | 605 | ||
NM_001278307.1 | NP_001265236.1 | 8 | 643 | ||
NM_001278308.1 | NP_001265237.1 | 9 | 676 | ||
NM_080678.2 | NP_542409.1 | 10 | 739 | ||
NR_103498.1 | 9 | 671 | |||
NR_103499.1 | 8 | 648 | |||
NR_103500.1 | 11 | 838 | |||
GenBank mRNA | AF310723.1 | - | 9 | 535 | |
AK293334.1 | - | 10 | 693 | ||
AK294107.1 | - | 9 | 551 | ||
AK297502.1 | - | 8 | 574 | ||
AK303094.1 | - | 8 | 597 | ||
BC010549.1 | - | 10 | 626 | 4153270 | |
BC039435.1 | - | 2 | 838 | ||
BM470609.1 | - | 469 | |||
BQ959565.1 | - | 9 | 538 |