Gene Transcripts

Gene Symbol
LYSMD4
Entrez Gene ID
145748
Gene Name
LysM domain containing 4
Gene Aliases
Chromosome Location
Chr. 15: 99715033 - 99733561 on Build GRCh38
UniGene ID
Hs.562568
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284417.1 NP_001271346.1 3 423
NM_001284418.1 NP_001271347.1 3 424
NM_001284419.1 NP_001271348.1 5 499
NM_001284420.1 NP_001271349.1 2 133
NM_001284421.1 NP_001271350.1 4 449
NM_001284422.1 NP_001271351.1 5 549
NM_152449.3 NP_689662.2 6 614
XM_006720397.3 XP_006720460.2 2 147
XM_011521241.1 XP_011519543.1 2 207
XM_011521242.2 XP_011519544.1 2 725
XM_011521243.2 XP_011519545.1 2 725
XM_011521244.1 XP_011519546.1 3 776
XM_011521245.2 XP_011519547.1 2 147
XM_017021926.1 XP_016877415.1 4 640
XM_017021927.1 XP_016877416.1 3 961
XM_017021928.1 XP_016877417.1 2 605
XM_017021929.1 XP_016877418.1 4 802
XM_017021930.1 XP_016877419.1 4 816
GenBank mRNA AK124852.1 - 4 752
AK290296.1 - 3 409
BC041097.1 - 6 591 5240362
BC084545.1 - 3 391 4590701
BG420645.1 - 3 379
CB989579.1 - 4 440
DA405564.1 - 5 492
DA594971.1 - 2 107
DB044672.1 - 5 499
HY131530.1 - 3 160

View Full Product Details