Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138477.2 | NP_612486.2 | 6 | 1199 | |
XM_005254176.4 | XP_005254233.1 | 6 | 1114 | ||
XM_011521270.2 | XP_011519572.1 | 6 | 1139 | ||
XM_011521271.2 | XP_011519573.1 | 6 | 1136 | ||
XM_011521274.2 | XP_011519576.1 | 3 | 185 | ||
GenBank mRNA | AF525398.1 | - | 6 | 1196 | |
BC052568.1 | - | 6 | 1087 | ||
BC066640.1 | - | 6 | 1087 | 6577335 | |
BC113821.1 | - | 4 | 283 |