Gene Transcripts

Gene Symbol
ZNF362
Entrez Gene ID
149076
Gene Name
zinc finger protein 362
Gene Aliases
RN, lin-29
Chromosome Location
Chr. 1: 33256307 - 33300719 on Build GRCh38
UniGene ID
Hs.743358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152493.2 NP_689706.2 2 183
XM_005270508.4 XP_005270565.1 1 863
XM_017000415.1 XP_016855904.1 1 895
GenBank mRNA AF318356.1 - 1 33
BC104779.1 - 1 105 8143782
BC112113.1 - 1 105 8327581
BX283663.1 - 2 184

View Full Product Details