Gene Transcripts

Gene Symbol
CNIH3
Entrez Gene ID
149111
Gene Name
cornichon family AMPA receptor auxiliary protein 3
Gene Aliases
CNIH-3
Chromosome Location
Chr. 1: 224434640 - 224740549 on Build GRCh38
UniGene ID
Hs.731723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322305.1 NP_001309234.1 1, 2 786
NM_152495.1 NP_689708.1 1, 2 786
NR_136287.1 1, 2 786
GenBank mRNA AK096312.1 - 1, 2 786
BC022780.1 - 1, 2 274 3894832

View Full Product Details