Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152512.3 | NP_689725.2 | 7 | 1549 | |
XM_006724149.3 | XP_006724212.1 | 7 | 2144 | ||
XM_006724150.3 | XP_006724213.1 | 7 | 1334 | ||
XM_006724151.1 | XP_006724214.1 | 6 | 1038 | ||
XM_011529927.2 | XP_011528229.1 | 7 | 1580 | ||
XM_011529928.2 | XP_011528230.1 | 6 | 1400 | ||
XM_011529930.2 | XP_011528232.1 | 3 | 565 | ||
GenBank mRNA | AK058150.1 | - | 2 | 231 | |
AK093154.1 | - | 5 | 783 | ||
BC033895.2 | - | 7 | 1549 | 4831324 | |
CR456452.1 | - | 6 | 1344 |