Gene Transcripts

Gene Symbol
ERICH1
Entrez Gene ID
157697
Gene Name
glutamate rich 1
Gene Aliases
HSPC319
Chromosome Location
Chr. 8: 614737 - 731239 on Build GRCh38
UniGene ID
Hs.655310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303100.1 NP_001290029.1 2 202
NM_207332.2 NP_997215.1 2 202
XM_006716234.3 XP_006716297.2 2 191
XM_011534731.2 XP_011533033.1 2 191
XM_011534735.2 XP_011533037.1 2 191
XM_011534736.2 XP_011533038.1 2 191
XM_011534737.2 XP_011533039.1 2 191
XM_011534739.2 XP_011533041.1 2 191
XM_017013114.1 XP_016868603.1 2 191
XM_017013115.1 XP_016868604.1 2 191
XM_017013116.1 XP_016868605.1 2 191
XM_017013117.1 XP_016868606.1 2 191
XM_017013118.1 XP_016868607.1 2 191
XM_017013119.1 XP_016868608.1 2 191
XM_017013120.1 XP_016868609.1 2 191
XM_017013121.1 XP_016868610.1 2 191
XM_017013122.1 XP_016868611.1 2 191
XM_017013123.1 XP_016868612.1 2 191
XM_017013124.1 XP_016868613.1 2 191
XM_017013125.1 XP_016868614.1 2 191
XM_017013126.1 XP_016868615.1 2 191
XM_017013127.1 XP_016868616.1 2 191
XM_017013128.1 XP_016868617.1 2 191
XM_017013129.1 XP_016868618.1 2 191
XM_017013130.1 XP_016868619.1 2 191
XM_017013131.1 XP_016868620.1 2 191
XM_017013132.1 XP_016868621.1 2 191
XM_017013133.1 XP_016868622.1 2 191
XM_017013134.1 XP_016868623.1 2 191
XM_017013135.1 XP_016868624.1 4 1743
XM_017013136.1 XP_016868625.1 4 1746
XM_017013137.1 XP_016868626.1 4 1776
XM_017013138.1 XP_016868627.1 4 1721
XM_017013139.1 XP_016868628.1 4 1718
GenBank mRNA AF161437.1 - 2 146
AK290264.1 - 2 186
AK297476.1 - 2 172
BC046243.1 - 2 189 5207311
BU540397.1 - 2 205

View Full Product Details