Gene Transcripts

Gene Symbol
CRTC2
Entrez Gene ID
200186
Gene Name
CREB regulated transcription coactivator 2
Gene Aliases
TORC-2, TORC2
Chromosome Location
Chr. 1: 153947672 - 153958656 on Build GRCh38
UniGene ID
Hs.406392
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181715.2 NP_859066.1 12 1749
XM_005244946.1 XP_005245003.1 12 1745
XM_005244949.2 XP_005245006.1 12 1622
XM_017000575.1 XP_016856064.1 11 1518
XM_017000576.1 XP_016856065.1 11 1503
XM_017000577.1 XP_016856066.1 11 1647
XM_017000578.1 XP_016856067.1 11 1632
XM_017000579.1 XP_016856068.1 6 1044
GenBank mRNA AF289574.1 - 3 708
AK096382.1 - 10 1340
AK223559.1 - 12 1702
AY360172.1 - 12 1720
AY726597.1 - 12 1529
BC033304.1 - 10 1351
BC053562.1 - 12 1660 6188068

View Full Product Details