Gene Transcripts
- Gene Symbol
- VWA3B
- Entrez Gene ID
- 200403
- Gene Name
- von Willebrand factor A domain containing 3B
- Gene Aliases
- SCAR22
- Chromosome Location
- Chr. 2: 98087132 - 98330675 on Build GRCh38
- UniGene ID
- Hs.269977
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144992.4 |
NP_659429.4
|
6, 7 |
1134 |
|
XM_005263897.2 |
XP_005263954.1
|
6, 7 |
1099 |
|
XM_006712357.2 |
XP_006712420.1
|
5, 6 |
848 |
|
XM_011510770.1 |
XP_011509072.1
|
6, 7 |
1099 |
|
XM_011510771.2 |
XP_011509073.1
|
6, 7 |
1099 |
|
XM_011510772.1 |
XP_011509074.1
|
6, 7 |
1099 |
|
XM_011510774.1 |
XP_011509076.1
|
6, 7 |
1099 |
|
XM_011510775.2 |
XP_011509077.1
|
6, 7 |
1099 |
|
XM_017003564.1 |
XP_016859053.1
|
6, 7 |
1099 |
|
GenBank mRNA |
AK292397.1 |
- |
6, 7 |
1081 |
|
AK302390.1 |
- |
5, 6 |
779 |
|
AL832635.2 |
- |
7, 8 |
1077 |
|
AL832761.1 |
- |
6, 7 |
1092 |
|
BC026091.2 |
- |
6, 7 |
1030 |
|
BC095480.1 |
- |
6, 7 |
1089 |
|
BC150653.1 |
- |
6, 7 |
1074 |
9057024 |
View Full Product Details