Gene Transcripts

Gene Symbol
ENKUR
Entrez Gene ID
219670
Gene Name
enkurin, TRPC channel interacting protein
Gene Aliases
C10orf63, CFAP106
Chromosome Location
Chr. 10: 24981979 - 25062279 on Build GRCh38
UniGene ID
Hs.534486
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145010.3 NP_659447.1 2 399
NR_072992.1 2 399
XM_006717389.1 XP_006717452.1 2 395
XM_011519348.1 XP_011517650.1 2 395
GenBank mRNA AK095021.1 - 2 344
AK292495.1 - 2 396
AY454125.1 - 2 386
BC026165.2 - 2 401 4837530

View Full Product Details