Gene Transcripts

Gene Symbol
PELI3
Entrez Gene ID
246330
Gene Name
pellino E3 ubiquitin protein ligase family member 3
Gene Aliases
Chromosome Location
Chr. 11: 66466327 - 66477337 on Build GRCh38
UniGene ID
Hs.656875
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098510.1 NP_001091980.1 6 789
NM_001243135.1 NP_001230064.1 6 628
NM_001243136.1 NP_001230065.1 5 636
NM_145065.2 NP_659502.2 7 861
XM_011544884.2 XP_011543186.1 7 864
XM_017017465.1 XP_016872954.1 5 613
GenBank mRNA AF487456.1 - 6 697
AF487457.1 - 5 625
AK074201.1 - 7 799
AK075253.1 - 3 301
AK094060.1 - 6 628
AL834395.1 - 7 808
BC021256.2 - 7 802 3535985
BC025723.1 - 6 710 5198999
BX341733.2 - 5 573

Gene Symbol
LOC101928069
Entrez Gene ID
101928069
Gene Name
uncharacterized LOC101928069
Gene Aliases
Chromosome Location
Chr. 11: 66473490 - 66480249 on Build GRCh38
UniGene ID
Hs.523816
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_120586.1 4 550
GenBank mRNA BQ223044.1 - 4 532

View Full Product Details