Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173566.2 | NP_775837.2 | 6 | 6112 | |
XM_006724219.3 | XP_006724282.1 | 6 | 6672 | ||
XM_006724220.3 | XP_006724283.1 | 6 | 6074 | ||
XM_006724221.2 | XP_006724284.1 | 5 | 5628 | ||
XM_011530080.2 | XP_011528382.1 | 7 | 6880 | ||
XM_011530081.2 | XP_011528383.1 | 6 | 6756 | ||
XM_017028736.1 | XP_016884225.1 | 6 | 7832 | ||
XM_017028737.1 | XP_016884226.1 | 5 | 7270 | ||
GenBank mRNA | AK123082.1 | - | 1 | 88 | |
AK130944.1 | - | 3 | 1926 | ||
BC040859.1 | - | 3 | 826 | 5535580 | |
BX647272.1 | - | 3 | 415 | ||
CT841517.1 | - | 5 | 5944 |