Gene Transcripts

Gene Symbol
CSNK1G2-AS1
Entrez Gene ID
255193
Gene Name
CSNK1G2 antisense RNA 1
Gene Aliases
C19orf34
Chromosome Location
Chr. 19: 1952527 - 1954549 on Build GRCh38
UniGene ID
Hs.374717
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033400.1 1, 2 697
GenBank mRNA BC029585.1 - 2, 3 641

View Full Product Details