Gene Transcripts

Gene Symbol
EML3
Entrez Gene ID
256364
Gene Name
echinoderm microtubule associated protein like 3
Gene Aliases
ELP95
Chromosome Location
Chr. 11: 62602218 - 62613020 on Build GRCh38
UniGene ID
Hs.379785
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300793.1 NP_001287722.1 7, 8 1238
NM_001300794.1 NP_001287723.1 7, 8 1235
NM_153265.2 NP_694997.2 7, 8 1235
XM_005273878.3 XP_005273935.1 7, 8 1493
XM_006718489.3 XP_006718552.1 7, 8 1610
XM_006718490.3 XP_006718553.1 7, 8 1610
XM_006718491.3 XP_006718554.1 7, 8 1607
XM_011544896.2 XP_011543198.1 7, 8 1607
XM_011544897.2 XP_011543199.1 7, 8 1610
XM_017017480.1 XP_016872969.1 7, 8 981
XM_017017481.1 XP_016872970.1 6, 7 867
XM_017017482.1 XP_016872971.1 7, 8 981
XM_017017483.1 XP_016872972.1 7, 8 1038
GenBank mRNA AK093146.1 - 6, 7 910
AK128679.1 - 6, 7 1652
AK296901.1 - 7, 8 1000
AK310579.1 - 9, 10 1061
AL833388.1 - 4, 5 490
BC075824.1 - 6, 7 841
BC108280.1 - 7, 8 1019 3915493

View Full Product Details