Gene Transcripts

Gene Symbol
ZSCAN1
Entrez Gene ID
284312
Gene Name
zinc finger and SCAN domain containing 1
Gene Aliases
MZF-1, ZNF915
Chromosome Location
Chr. 19: 58033989 - 58058907 on Build GRCh38
UniGene ID
Hs.643437
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182572.3 NP_872378.3 6 970
XM_006723149.2 XP_006723212.1 5 2583
GenBank mRNA AK091098.1 - 6 970

View Full Product Details