Gene Transcripts

Gene Symbol
STPG2
Entrez Gene ID
285555
Gene Name
sperm tail PG-rich repeat containing 2
Gene Aliases
C4orf37
Chromosome Location
Chr. 4: 97441450 - 98143473 on Build GRCh38
UniGene ID
Hs.680226
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174952.2 NP_777612.1 7 912
XM_011531886.2 XP_011530188.1 9 1412
XM_017008049.1 XP_016863538.1 7 1187
XM_017008050.1 XP_016863539.1 7 1145
XM_017008051.1 XP_016863540.1 4 390
GenBank mRNA BC036870.1 - 7 912

View Full Product Details