Gene Transcripts
- Gene Symbol
- NRBP2
- Entrez Gene ID
- 340371
- Gene Name
- nuclear receptor binding protein 2
- Gene Aliases
- TRG16, pp9320
- Chromosome Location
- Chr. 8: 143829776 - 143840974 on Build GRCh38
- UniGene ID
- Hs.521926
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178564.3 |
NP_848659.2
|
10, 11 |
979 |
|
XM_017013378.1 |
XP_016868867.1
|
10, 11 |
979 |
|
XM_017013379.1 |
XP_016868868.1
|
10, 11 |
979 |
|
XM_017013380.1 |
XP_016868869.1
|
10, 11 |
979 |
|
XM_017013381.1 |
XP_016868870.1
|
10, 11 |
979 |
|
XM_017013382.1 |
XP_016868871.1
|
10, 11 |
979 |
|
XM_017013383.1 |
XP_016868872.1
|
10, 11 |
979 |
|
GenBank mRNA |
AK293943.1 |
- |
10, 11 |
979 |
|
AL137662.1 |
- |
8, 9 |
908 |
|
AL834530.1 |
- |
3, 4 |
218 |
|
BC071605.1 |
- |
9, 10 |
1142 |
|
BC113873.1 |
- |
7, 8 |
983 |
40053695 |
BC114481.1 |
- |
8, 9 |
880 |
|
CR457350.1 |
- |
2, 3 |
110 |
|
View Full Product Details