Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304506.1 | NP_001291435.1 | 5 | 1063 | |
NM_019110.4 | NP_061983.2 | 5 | 1728 | ||
XM_005249095.3 | XP_005249152.1 | 7 | 1772 | ||
XM_005249097.3 | XP_005249154.1 | 7 | 1468 | ||
XM_005249098.3 | XP_005249155.1 | 5 | 1386 | ||
XM_005249100.4 | XP_005249157.2 | 5 | 1119 | ||
XM_006715095.3 | XP_006715158.1 | 6 | 1245 | ||
XM_011514588.2 | XP_011512890.1 | 5 | 1628 | ||
XM_011514589.2 | XP_011512891.1 | 5 | 1184 | ||
XM_017010844.1 | XP_016866333.1 | 7 | 2423 | ||
GenBank mRNA | AK302017.1 | - | 5 | 1044 | |
AK315783.1 | - | 5 | 1698 | ||
AY781778.1 | - | 5 | 1698 | ||
BC014031.2 | - | 5 | 1656 | 4336904 | |
KU178905.1 | - | 5 | 1412 | ||
KU178906.1 | - | 4 | 1301 |