Gene Transcripts

Gene Symbol
RNASE9
Entrez Gene ID
390443
Gene Name
ribonuclease A family member 9 (inactive)
Gene Aliases
HEL128, RAK1, h461
Chromosome Location
Chr. 14: 20556093 - 20560931 on Build GRCh38
UniGene ID
Hs.533972
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001673.3 NP_001001673.2 2 533
NM_001110356.1 NP_001103826.1 3 585
NM_001110357.1 NP_001103827.1 3 568
NM_001110358.1 NP_001103828.1 4 621
NM_001110359.1 NP_001103829.1 5 731
NM_001110360.1 NP_001103830.1 4 696
NM_001110361.1 NP_001103831.1 5 656
NM_001289110.1 NP_001276039.1 4 620
GenBank mRNA AF382949.1 - 1 461
AY665804.1 - 1 461
AY907670.1 - 2 533
BC130311.1 - 1 546 40146342
BC130313.1 - 1 546 40146344
EF061296.1 - 3 568
EF061297.1 - 3 585
EF061298.1 - 4 620
EF061299.1 - 4 621
EF061300.1 - 4 696
EF061301.1 - 5 656
EF061302.1 - 5 731
EF061303.1 - 3 2152
EF061304.1 - 4 2187
EU414264.1 - 1 461

View Full Product Details