Gene Transcripts

Gene Symbol
SLC25A35
Entrez Gene ID
399512
Gene Name
solute carrier family 25 member 35
Gene Aliases
Chromosome Location
Chr. 17: 8287763 - 8295694 on Build GRCh38
UniGene ID
Hs.118918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320870.1 NP_001307799.1 1 752
NM_001320871.1 NP_001307800.1 1 752
NM_001320872.1 NP_001307801.1 1 752
NM_001320877.1 NP_001307806.1 1 752
NM_201520.2 NP_958928.1 1 752
NR_135483.1 1 752
NR_135484.1 1 752
XM_005256641.2 XP_005256698.1 1 809
XM_011523846.1 XP_011522148.1 1 809
XM_017024633.1 XP_016880122.1 1 809
XM_017024634.1 XP_016880123.1 1 1103
GenBank mRNA AK097536.1 - 1 261
AY498866.1 - 1 216
BC041597.1 - 1 752
BC052233.1 - 1 752
BC101329.2 - 1 247 40025001
BC101330.2 - 1 245 40025003
BC105995.1 - 1 727 4746077

View Full Product Details