Gene Transcripts

Gene Symbol
FSIP2
Entrez Gene ID
401024
Gene Name
fibrous sheath interacting protein 2
Gene Aliases
Chromosome Location
Chr. 2: 185738895 - 185833289 on Build GRCh38
UniGene ID
Hs.98025
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173651.3 NP_775922.3 17 18733
GenBank mRNA AK126036.1 - 1 1050
AK126051.1 - 1 2239
BX647691.1 - 2 1829
BX648733.1 - 1 3006

View Full Product Details