Gene Transcripts

Gene Symbol
PEAK1
Entrez Gene ID
79834
Gene Name
pseudopodium enriched atypical kinase 1
Gene Aliases
SGK269
Chromosome Location
Chr. 15: 77100829 - 77420104 on Build GRCh38
UniGene ID
Hs.731402
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024776.3 NP_079052.2 7 4046
XM_005254670.2 XP_005254727.1 10 4350
XM_005254671.2 XP_005254728.1 9 4247
XM_005254673.2 XP_005254730.1 7 3981
XM_005254674.2 XP_005254731.1 8 4194
XM_005254675.4 XP_005254732.1 4 3700
XM_011522033.2 XP_011520335.1 11 4406
XM_011522034.2 XP_011520336.1 12 4514
XM_011522035.2 XP_011520337.1 14 4665
XM_011522036.2 XP_011520338.1 13 4569
XM_011522037.2 XP_011520339.1 13 4610
XM_011522038.2 XP_011520340.1 12 4461
XM_011522039.2 XP_011520341.1 13 4557
XM_011522040.1 XP_011520342.1 11 4406
XM_011522042.2 XP_011520344.1 3 783
GenBank mRNA AB082533.1 - 3 525
AK127658.1 - 3 1508

View Full Product Details