Gene Transcripts
- Gene Symbol
- CAMK2B
- Entrez Gene ID
- 816
- Gene Name
- calcium/calmodulin dependent protein kinase II beta
- Gene Aliases
- CAM2, CAMK2, CAMKB
- Chromosome Location
- Chr. 7: 44217150 - 44325994 on Build GRCh38
- UniGene ID
- Hs.351887
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001220.4 |
NP_001211.3
|
4, 5 |
474 |
|
NM_001293170.1 |
NP_001280099.1
|
4, 5 |
474 |
|
NM_172078.2 |
NP_742075.1
|
4, 5 |
474 |
|
NM_172079.2 |
NP_742076.1
|
4, 5 |
474 |
|
NM_172080.2 |
NP_742077.1
|
4, 5 |
474 |
|
NM_172081.2 |
NP_742078.1
|
4, 5 |
474 |
|
NM_172082.2 |
NP_742079.1
|
4, 5 |
474 |
|
NM_172083.2 |
NP_742080.1
|
4, 5 |
474 |
|
NM_172084.2 |
NP_742081.1
|
4, 5 |
474 |
|
XM_005249862.3 |
XP_005249919.1
|
4, 5 |
350 |
|
XM_005249864.3 |
XP_005249921.1
|
4, 5 |
350 |
|
XM_006715781.2 |
XP_006715844.1
|
4, 5 |
350 |
|
XM_006715784.2 |
XP_006715847.1
|
4, 5 |
350 |
|
XM_011515547.1 |
XP_011513849.1
|
4, 5 |
350 |
|
XM_011515549.1 |
XP_011513851.1
|
4, 5 |
350 |
|
XM_011515550.1 |
XP_011513852.1
|
4, 5 |
350 |
|
XM_011515551.1 |
XP_011513853.1
|
4, 5 |
350 |
|
XM_011515552.1 |
XP_011513854.1
|
4, 5 |
350 |
|
XM_011515553.1 |
XP_011513855.1
|
4, 5 |
350 |
|
XM_011515554.1 |
XP_011513856.1
|
4, 5 |
350 |
|
XM_011515555.1 |
XP_011513857.1
|
4, 5 |
350 |
|
XM_011515556.1 |
XP_011513858.1
|
4, 5 |
350 |
|
XM_011515557.1 |
XP_011513859.1
|
4, 5 |
350 |
|
XM_011515558.2 |
XP_011513860.1
|
4, 5 |
350 |
|
XM_011515559.1 |
XP_011513861.1
|
4, 5 |
259 |
|
XM_017012660.1 |
XP_016868149.1
|
4, 5 |
350 |
|
XM_017012661.1 |
XP_016868150.1
|
4, 5 |
350 |
|
XM_017012662.1 |
XP_016868151.1
|
4, 5 |
350 |
|
XM_017012663.1 |
XP_016868152.1
|
4, 5 |
259 |
|
XM_017012664.1 |
XP_016868153.1
|
4, 5 |
350 |
|
XM_017012665.1 |
XP_016868154.1
|
4, 5 |
350 |
|
XM_017012666.1 |
XP_016868155.1
|
4, 5 |
259 |
|
XM_017012667.1 |
XP_016868156.1
|
4, 5 |
259 |
|
XM_017012668.1 |
XP_016868157.1
|
4, 5 |
259 |
|
XM_017012669.1 |
XP_016868158.1
|
4, 5 |
259 |
|
XM_017012670.1 |
XP_016868159.1
|
4, 5 |
259 |
|
GenBank mRNA |
AF078803.1 |
- |
4, 5 |
267 |
|
AF081572.1 |
- |
4, 5 |
267 |
|
AF081924.1 |
- |
4, 5 |
267 |
|
AF083419.1 |
- |
4, 5 |
267 |
|
AF112471.1 |
- |
4, 5 |
310 |
|
AF112472.1 |
- |
4, 5 |
310 |
|
AF140350.1 |
- |
4, 5 |
267 |
|
AJ252236.1 |
- |
4, 5 |
351 |
|
AJ252237.1 |
- |
4, 5 |
274 |
|
AJ252238.1 |
- |
4, 5 |
264 |
|
AK222703.1 |
- |
4, 5 |
298 |
|
AK290148.1 |
- |
4, 5 |
339 |
|
AK315663.1 |
- |
4, 5 |
381 |
|
BC019070.2 |
- |
4, 5 |
468 |
5014712 |
BI550751.1 |
- |
|
479 |
|
BX445032.2 |
- |
4, 5 |
283 |
|
DA069361.1 |
- |
3, 4 |
126 |
|
DB175099.1 |
- |
4, 5 |
252 |
|
U23460.1 |
- |
4, 5 |
339 |
|
View Full Product Details