Gene Transcripts

Gene Symbol
DUSP2
Entrez Gene ID
1844
Gene Name
dual specificity phosphatase 2
Gene Aliases
PAC-1, PAC1
Chromosome Location
Chr. 2: 96143166 - 96145468 on Build GRCh38
UniGene ID
Hs.1183
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004418.3 NP_004409.1 2 506
XM_017003546.1 XP_016859035.1 1 618
GenBank mRNA BC007771.2 - 2 482
L11329.1 - 2 505

View Full Product Details