Gene Transcripts

Gene Symbol
CDX1
Entrez Gene ID
1044
Gene Name
caudal type homeobox 1
Gene Aliases
Chromosome Location
Chr. 5: 150166781 - 150184558 on Build GRCh38
UniGene ID
Hs.1545
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001804.2 NP_001795.2 3 730
GenBank mRNA AF239666.1 - 3 715
BC096250.1 - 3 200
BC096251.1 - 4 347
BC096252.1 - 4 393 40005955
BC127122.1 - 4 393 40005957
U15212.1 - 3 448
U51095.1 - 3 715

View Full Product Details