Gene Transcripts

Gene Symbol
CRYBB1
Entrez Gene ID
1414
Gene Name
crystallin beta B1
Gene Aliases
CATCN3, CTRCT17
Chromosome Location
Chr. 22: 26599278 - 26618103 on Build GRCh38
UniGene ID
Hs.37135
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001887.3 NP_001878.1 2 234
XM_011529899.2 XP_011528201.1 2 300
GenBank mRNA BC036790.1 - 2 281
CR456425.1 - 2 203
U35340.1 - 2 236

View Full Product Details