Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001899.2 | NP_001890.1 | 1 | 82 | |
GenBank mRNA | BC065714.1 | - | 1 | 99 | |
BC074952.2 | - | 1 | 55 | 30915278 | |
BC074953.2 | - | 1 | 55 | 30915500 | |
X54667.1 | - | 1 | 82 |