Gene Transcripts

Gene Symbol
DMWD
Entrez Gene ID
1762
Gene Name
dystrophia myotonica, WD repeat containing
Gene Aliases
D19S593E, DMR-N9, DMRN9, gene59
Chromosome Location
Chr. 19: 45783006 - 45792802 on Build GRCh38
UniGene ID
Hs.515474
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004943.1 NP_004934.1 1 430
GenBank mRNA BM922103.1 - 430
L19267.1 - 1 18

View Full Product Details