Gene Transcripts

Gene Symbol
FOXG1
Entrez Gene ID
2290
Gene Name
forkhead box G1
Gene Aliases
BF1, BF2, FHKL3, FKH2, FKHL1, FKHL2, FKHL3, FKHL4, FOXG1A, FOXG1B, FOXG1C, HBF-1, HBF-2, HBF-3, HBF-G2, HBF2, HFK1, HFK2, HFK3, KHL2, QIN
Chromosome Location
Chr. 14: 28767072 - 28770277 on Build GRCh38
UniGene ID
Hs.632336
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005249.4 NP_005240.3 1 1236
GenBank mRNA BC050072.1 - 2 1397

View Full Product Details