Gene Transcripts

Gene Symbol
MYL4
Entrez Gene ID
4635
Gene Name
myosin light chain 4
Gene Aliases
ALC1, AMLC, GT1, PRO1957
Chromosome Location
Chr. 17: 47209017 - 47227650 on Build GRCh38
UniGene ID
Hs.463300
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002841.1 NP_001002841.1 5 473
NM_002476.2 NP_002467.1 4 420
XM_005257391.4 XP_005257448.1 5 400
XM_011524839.2 XP_011523141.2 5 560
XM_017024683.1 XP_016880172.1 5 547
XM_017024684.1 XP_016880173.1 4 564
GenBank mRNA AF116676.2 - 772
BC030228.1 - 4 409 5226518
BU658678.1 - 5 479
M24121.1 - 4 407
M36172.1 - 4 401
X13955.1 - 4 401
X52005.1 - 157

View Full Product Details