Gene Transcripts
- Gene Symbol
- NME4
- Entrez Gene ID
- 4833
- Gene Name
- NME/NM23 nucleoside diphosphate kinase 4
- Gene Aliases
- NDPK-D, NM23H4, nm23-H4
- Chromosome Location
- Chr. 16: 396755 - 400754 on Build GRCh38
- UniGene ID
- Hs.9235
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286435.1 |
NP_001273364.1
|
2, 3 |
251 |
|
NM_001286436.1 |
NP_001273365.1
|
2, 3 |
454 |
|
NM_001286438.1 |
NP_001273367.1
|
3, 4 |
429 |
|
NM_001286439.1 |
NP_001273368.1
|
3, 4 |
438 |
|
NM_001286440.1 |
NP_001273369.1
|
2, 3 |
567 |
|
NM_005009.2 |
NP_005000.1
|
2, 3 |
251 |
|
GenBank mRNA |
BC004880.2 |
- |
2, 3 |
234 |
|
BC017067.1 |
- |
2, 3 |
251 |
3913604 |
BE266496.1 |
- |
2, 3 |
206 |
|
BP328820.1 |
- |
3, 4 |
324 |
|
BQ650649.1 |
- |
2, 3 |
410 |
|
BQ932845.1 |
- |
2, 3 |
505 |
|
BT019438.1 |
- |
2, 3 |
220 |
|
CB143020.1 |
- |
2, 3 |
454 |
|
CF126980.1 |
- |
3, 4 |
403 |
|
Y07604.1 |
- |
2, 3 |
231 |
|
View Full Product Details