Gene Transcripts
- Gene Symbol
- REG1B
- Entrez Gene ID
- 5968
- Gene Name
- regenerating family member 1 beta
- Gene Aliases
- PSPS2, REGH, REGI-BETA, REGL
- Chromosome Location
- Chr. 2: 79085023 - 79088024 on Build GRCh38
- UniGene ID
- Hs.4158
- Species
- Homo sapiens
- Gene Symbol
- NKX2-2
- Entrez Gene ID
- 4821
- Gene Name
- NK2 homeobox 2
- Gene Aliases
- NKX2.2, NKX2B
- Chromosome Location
- Chr. 20: 21511010 - 21514026 on Build GRCh38
- UniGene ID
- Hs.516922
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BM314196.1 |
- |
|
254 |
|
View Full Product Details