Gene Transcripts

Gene Symbol
SRL
Entrez Gene ID
6345
Gene Name
sarcalumenin
Gene Aliases
Chromosome Location
Chr. 16: 4189374 - 4242253 on Build GRCh38
UniGene ID
Hs.10041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098814.1 NP_001092284.1 3, 4 263
NM_001323667.1 NP_001310596.1 4, 5 393
NM_001323668.1 NP_001310597.1 4, 5 1641
XM_017023527.1 XP_016879016.1 4, 5 1588
XM_017023528.1 XP_016879017.1 3, 4 294
XM_017023529.1 XP_016879018.1 3, 4 309
GenBank mRNA AK056588.1 - 3, 4 263
AK302601.1 - 3, 4 299
AL831973.2 - 3, 4 235
AL832361.1 - 4, 5 1635
BX647476.1 - 4, 5 1635
DB149163.1 - 3, 4 263

View Full Product Details