Gene Transcripts

Gene Symbol
UBE2G2
Entrez Gene ID
7327
Gene Name
ubiquitin conjugating enzyme E2 G2
Gene Aliases
UBC7
Chromosome Location
Chr. 21: 44768580 - 44801836 on Build GRCh38
UniGene ID
Hs.529420
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001202489.1 NP_001189418.1 3, 4 385
NM_003343.5 NP_003334.2 5, 6 467
NM_182688.2 NP_872630.1 6, 7 582
XM_017028469.1 XP_016883958.1 5, 6 426
GenBank mRNA AF032456.1 - 5, 6 434
AK075143.1 - 1, 2 608
AK122700.1 - 1, 2 1884
AK290629.1 - 5, 6 467
AL834129.1 - 1, 2 1199
BC001738.1 - 5, 6 429 3536853
BC008351.1 - 5, 6 427 3536171
BC011569.1 - 5, 6 439 4548763
BE292782.1 - 6, 7 521
BG702453.1 - 5, 6 458
BT006914.1 - 5, 6 379
DA710059.1 - 3, 4 376

View Full Product Details