Gene Transcripts
- Gene Symbol
- HIST1H3J
- Entrez Gene ID
- 8356
- Gene Name
- histone cluster 1, H3j
- Gene Aliases
- H3/j, H3FJ
- Chromosome Location
- Chr. 6: 27890315 - 27890792 on Build GRCh38
- UniGene ID
- Hs.484990
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003535.2 |
NP_003526.1
|
1 |
397 |
|
View Full Product Details