Gene Transcripts

Gene Symbol
H1FX
Entrez Gene ID
8971
Gene Name
H1 histone family member X
Gene Aliases
H1.10, H1X
Chromosome Location
Chr. 3: 129314771 - 129316277 on Build GRCh38
UniGene ID
Hs.75307
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006026.3 NP_006017.1 1 624
GenBank mRNA BC000426.2 - 1 624
BC010435.1 - 1 330 3538400
D64142.1 - 1 350

View Full Product Details