Gene Transcripts
- Gene Symbol
- ATP6V1F
- Entrez Gene ID
- 9296
- Gene Name
- ATPase H+ transporting V1 subunit F
- Gene Aliases
- ATP6S14, VATF, Vma7
- Chromosome Location
- Chr. 7: 128862803 - 128865849 on Build GRCh38
- UniGene ID
- Hs.78089
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001198909.1 |
NP_001185838.1
|
2, 3 |
354 |
|
NM_004231.3 |
NP_004222.2
|
1, 2 |
270 |
|
GenBank mRNA |
AK312214.1 |
- |
1, 2 |
215 |
|
BC104230.1 |
- |
1, 2 |
221 |
40032234 |
BC104231.1 |
- |
1, 2 |
221 |
40032235 |
BC107854.1 |
- |
1, 2 |
247 |
5171984 |
BG724326.1 |
- |
1, 2 |
255 |
|
BU683155.1 |
- |
1 |
475 |
|
BU956696.1 |
- |
2, 3 |
253 |
|
CR456896.1 |
- |
1, 2 |
168 |
|
D49400.1 |
- |
1, 2 |
218 |
|
DB499437.1 |
- |
1, 2 |
270 |
|
View Full Product Details