Gene Transcripts

Gene Symbol
KBTBD11
Entrez Gene ID
9920
Gene Name
kelch repeat and BTB domain containing 11
Gene Aliases
KLHDC7C
Chromosome Location
Chr. 8: 1970718 - 2006943 on Build GRCh38
UniGene ID
Hs.5333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014867.2 NP_055682.1 2 2456
XM_011534771.2 XP_011533073.1 2 3126
XM_011534772.2 XP_011533074.1 3 2525
XM_017014114.1 XP_016869603.1 4 3060
XM_017014115.1 XP_016869604.1 3 2914
XM_017014116.1 XP_016869605.1 3 2581
XM_017014117.1 XP_016869606.1 2 2845
GenBank mRNA AB018254.1 - 2 2453
AY762229.1 - 1 1490

View Full Product Details