Gene Transcripts
- Gene Symbol
- SCAMP2
- Entrez Gene ID
- 10066
- Gene Name
- secretory carrier membrane protein 2
- Gene Aliases
- Chromosome Location
- Chr. 15: 74843730 - 74873379 on Build GRCh38
- UniGene ID
- Hs.458917
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320778.1 |
NP_001307707.1
|
6, 7 |
730 |
|
NM_005697.4 |
NP_005688.2
|
5, 6 |
601 |
|
XM_017021856.1 |
XP_016877345.1
|
6, 7 |
655 |
|
GenBank mRNA |
AF005038.2 |
- |
5, 6 |
506 |
|
AK096376.1 |
- |
4, 5 |
492 |
|
AK096794.1 |
- |
4, 5 |
2390 |
|
AK291884.1 |
- |
5, 6 |
587 |
|
AK315516.1 |
- |
5, 6 |
550 |
|
BC001376.2 |
- |
5, 6 |
551 |
3050527 |
BC004385.1 |
- |
5, 6 |
551 |
3617954 |
CD300387.1 |
- |
6, 7 |
706 |
|
CR456800.1 |
- |
5, 6 |
477 |
|
View Full Product Details