Gene Transcripts

Gene Symbol
GAS2L1
Entrez Gene ID
10634
Gene Name
growth arrest specific 2 like 1
Gene Aliases
GAR22
Chromosome Location
Chr. 22: 29306618 - 29312789 on Build GRCh38
UniGene ID
Hs.322852
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278730.1 NP_001265659.1 2 364
NM_006478.4 NP_006469.2 2 392
NM_152236.2 NP_689422.1 2 397
XM_011529824.1 XP_011528126.1 3 516
XM_011529825.1 XP_011528127.1 2 325
XM_017028533.1 XP_016884022.1 2 1042
GenBank mRNA AK093386.1 - 2 334
AK122829.1 - 1 1234
AK223602.1 - 2 336
AK225484.1 - 2 336
BC001782.1 - 2 332 3355168
BC007624.1 - 2 332 3357387
BC011047.1 - 2 335 4178571
Y07846.1 - 2 318

View Full Product Details