Gene Transcripts

Gene Symbol
PTTG2
Entrez Gene ID
10744
Gene Name
pituitary tumor-transforming 2
Gene Aliases
Chromosome Location
Chr. 4: 37960435 - 37961010 on Build GRCh38
UniGene ID
Hs.668806
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006607.2 NP_006598.2 1 360
GenBank mRNA AF095288.1 - 1 360
BC069114.1 - 1 360 7216932
BC069400.1 - 1 360 7262134

View Full Product Details