Gene Transcripts

Gene Symbol
GADD45G
Entrez Gene ID
10912
Gene Name
growth arrest and DNA damage inducible gamma
Gene Aliases
CR6, DDIT2, GADD45gamma, GRP17
Chromosome Location
Chr. 9: 89605009 - 89606555 on Build GRCh38
UniGene ID
Hs.9701
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006705.3 NP_006696.1 4 560
XM_011518163.2 XP_011516465.1 3 461
GenBank mRNA AF078078.1 - 4 465
AF079806.1 - 4 547
AF087883.1 - 4 545
AK313689.1 - 4 558
BC000465.2 - 4 541 2822765
BC019325.1 - 4 541 2823131
BT007234.1 - 4 450
CR457038.1 - 4 450
D83023.1 - 4 533

View Full Product Details